2-Bromo-3-methyl-5-(trimethylsilyl)thiophene ≥95%

Supplier: AOBChem USA

Warning

168807-1G 168807-500MG 168807-5G 168807-25G
77465-754EA 415.09 USD
77465-754 77478-510 77488-702 77499-222
2-Bromo-3-methyl-5-(trimethylsilyl)thiophene ≥95%
2-Bromo-3-methyl-5-(trimethylsilyl)thiophene

Precaución: For research use only

Formula: C₈H₁₃BrSSi
MW: 249.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34852491
CAS Number: 1448781-49-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR