α-Methyl-3-(1-methylethyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

168698-1G 168698-500MG 168698-5G 168698-25G
77465-742EA 314.15 USD
77465-742 77478-498 77488-690 77499-220
α-Methyl-3-(1-methylethyl)benzenemethanol ≥95%
α-Methyl-3-(1-methylethyl)benzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₆O
MW: 164.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11520980
CAS Number: 99723-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR