α,α,3,5-Tetramethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

168688-1G 168688-500MG 168688-5G
77465-738EA 534.57 USD
77465-738 77478-494 77488-686
α,α,3,5-Tetramethylbenzenemethanol ≥95%
α,α,3,5-Tetramethylbenzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD09965925
CAS Number: 34696-74-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR