m-Cymen-8-ol ≥95%

Supplier: AOBChem USA

Warning

168657-1G 168657-500MG 168657-5G
77465-734EA 552.08 USD
77465-734 77478-490 77488-680
m-Cymen-8-ol ≥95%
m-Cymen-8-ol

Precaución: For research use only

Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12067996
CAS Number: 5208-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR