4-(Dimethylamino)-α,α-dimethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

168645-1G 168645-500MG 168645-5G 168645-25G
77465-730EA 341.96 USD
77465-730 77478-486 77488-676 77499-210
4-(Dimethylamino)-α,α-dimethylbenzenemethanol ≥95%
4-(Dimethylamino)-α,α-dimethylbenzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₇NO
Storage Temperature: Ambient
MDL Number: MFCD20528514
CAS Number: 83026-55-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR