α,α-Dimethyl-4-propylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

168633-1G 168633-500MG 168633-5G
77465-726EA 471.74 USD
77465-726 77478-482 77488-672
α,α-Dimethyl-4-propylbenzenemethanol ≥95%
α,α-Dimethyl-4-propylbenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₈O
Storage Temperature: Ambient
MDL Number: MFCD17223765
CAS Number: 1267664-91-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR