3-(4-Bromo-2-fluorophenyl)-1-propene ≥95%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
4-Bromo-2-fluoro-1-(2-propen-1-yl)benzene
168274-1G
168274-500MG
168274-5G
168274-25G
77465-718EA
320.33
USD
77465-718
77478-474
77488-662
77499-198
3-(4-Bromo-2-fluorophenyl)-1-propene ≥95%
3-(4-Bromo-2-fluorophenyl)-1-propene
Precaución: For research use only
Formula:
C₉H₈BrF MW: 215.07 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09801298 CAS Number: 951894-09-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...