1-(Ethylthio)-3-(1-methylethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

168092-1G 168092-500MG 168092-5G
77465-714EA 648.9 USD
77465-714 77478-470 77488-658
1-(Ethylthio)-3-(1-methylethoxy)benzene ≥95%
1-(Ethylthio)-3-(1-methylethoxy)benzene

Precaución: For research use only

Formula: C₁₁H₁₆OS
MW: 196.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11617708
CAS Number: 1314950-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR