4-Ethoxy-1-(ethylthio)-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

168030-1G 168030-500MG 168030-5G 168030-25G
77465-710EA 415.09 USD
77465-710 77478-466 77488-654 77499-194
4-Ethoxy-1-(ethylthio)-2-methylbenzene ≥95%
4-Ethoxy-1-(ethylthio)-2-methylbenzene

Precaución: For research use only

Formula: C₁₁H₁₆OS
MW: 196.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34852414
CAS Number: 3765-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR