2-(Ethylthio)-4-methoxy-1-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

168026-1G 168026-500MG 168026-5G 168026-25G
77465-706EA 415.09 USD
77465-706 77478-462 77488-650 77499-192
2-(Ethylthio)-4-methoxy-1-methylbenzene ≥95%
2-(Ethylthio)-4-methoxy-1-methylbenzene

Precaución: For research use only

Formula: C₁₀H₁₄OS
Storage Temperature: Ambient
MDL Number: MFCD22483969
CAS Number: 85428-48-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR