1-(1,1-Dimethylethyl)-3-(ethylthio)benzene ≥95%

Supplier: AOBChem USA

Warning

168024-1G 168024-500MG 168024-5G
77465-702EA 485.13 USD
77465-702 77478-458 77488-646
1-(1,1-Dimethylethyl)-3-(ethylthio)benzene ≥95%
1-(1,1-Dimethylethyl)-3-(ethylthio)benzene

Precaución: For research use only

Formula: C₁₂H₁₈S
MW: 194.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11617719
CAS Number: 88315-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR