1-(Ethylthio)-3,5-dimethoxybenzene ≥95%

Supplier: AOBChem USA

Warning

168010-1G 168010-500MG 168010-5G
77465-698EA 668.47 USD
77465-698 77478-454 77488-642
1-(Ethylthio)-3,5-dimethoxybenzene ≥95%
1-(Ethylthio)-3,5-dimethoxybenzene

Precaución: For research use only

Formula: C₁₀H₁₄O₂S
MW: 198.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09030450
CAS Number: 849919-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR