3-(Ethylthio)-N,N-dimethylbenzenamine ≥95%

Supplier: AOBChem USA

Warning

167988-1G 167988-500MG 167988-5G 167988-25G
77465-694EA 415.09 USD
77465-694 77478-450 77488-638 77499-186
3-(Ethylthio)-N,N-dimethylbenzenamine ≥95%
3-(Ethylthio)-N,N-dimethylbenzenamine

Precaución: For research use only

Formula: C₁₀H₁₅NS
MW: 181.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34852407
CAS Number: 91330-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR