2-(Ethylthio)-1,4-dimethylbenzene ≥95%

Supplier: AOBChem USA

Warning

167983-1G 167983-500MG 167983-5G
77465-690EA 648.9 USD
77465-690 77478-446 77488-634
2-(Ethylthio)-1,4-dimethylbenzene ≥95%
2-(Ethylthio)-1,4-dimethylbenzene

Precaución: For research use only

Formula: C₁₀H₁₄S
MW: 166.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11617738
CAS Number: 158321-96-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR