4-(Ethylthio)-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

167959-1G 167959-500MG 167959-5G
77465-686EA 648.9 USD
77465-686 77478-442 77488-630
4-(Ethylthio)-1,1'-biphenyl ≥95%
4-(Ethylthio)-1,1'-biphenyl

Precaución: For research use only

Formula: C₁₄H₁₄S
MW: 214.33 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11521264
CAS Number: 100887-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR