1-Ethoxy-4-(ethylthio)benzene ≥95%

Supplier: AOBChem USA

Warning

167954-1G 167954-500MG 167954-5G
77465-682EA 668.47 USD
77465-682 77478-438 77488-626
1-Ethoxy-4-(ethylthio)benzene ≥95%
1-Ethoxy-4-(ethylthio)benzene

Precaución: For research use only

Formula: C₁₀H₁₄OS
Storage Temperature: Ambient
MDL Number: MFCD11617732
CAS Number: 82961-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR