1-(Ethylthio)-2,4-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

167948-1G 167948-500MG 167948-5G 167948-25G
77465-678EA 339.9 USD
77465-678 77478-434 77488-622 77499-182
1-(Ethylthio)-2,4-dimethylbenzene ≥97%
1-(Ethylthio)-2,4-dimethylbenzene

Precaución: For research use only

Formula: C₁₀H₁₄S
MW: 166.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11617694
CAS Number: 107832-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR