2-(Ethylthio)-1,4-dimethoxybenzene ≥95%

Supplier: AOBChem USA

Warning

167944-1G 167944-500MG 167944-5G
77465-674EA 648.9 USD
77465-674 77478-430 77488-618
2-(Ethylthio)-1,4-dimethoxybenzene ≥95%
2-(Ethylthio)-1,4-dimethoxybenzene

Precaución: For research use only

Formula: C₁₀H₁₄O₂S
MW: 198.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09030437
CAS Number: 24920-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR