1-(Ethylthio)naphthalene ≥95%

Supplier: AOBChem USA

Warning

167936-1G 167936-500MG 167936-5G
77465-670EA 648.9 USD
77465-670 77478-426 77488-614
1-(Ethylthio)naphthalene ≥95%
1-(Ethylthio)naphthalene

Precaución: For research use only

Formula: C₁₂H₁₂S
Storage Temperature: Ambient
MDL Number: MFCD11521263
CAS Number: 17539-31-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR