1-(Ethylthio)-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

167934-1G 167934-500MG 167934-5G
77465-666EA 668.47 USD
77465-666 77478-422 77488-610
1-(Ethylthio)-2-methylbenzene ≥95%
1-(Ethylthio)-2-methylbenzene

Precaución: For research use only

Formula: C₉H₁₂S
MW: 152.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11617743
CAS Number: 3695-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR