Ethyl 4-(1,1-dimethylpropyl)-α-oxobenzeneacetate ≥95%

Supplier: AOBChem USA

Warning

167811-1G 167811-500MG 167811-5G 167811-25G
77465-662EA 415.09 USD
77465-662 77478-418 77488-606 77499-174
Ethyl 4-(1,1-dimethylpropyl)-α-oxobenzeneacetate ≥95%
Ethyl 4-(1,1-dimethylpropyl)-α-oxobenzeneacetate

Precaución: For research use only

Formula: C₁₅H₂₀O₃
MW: 248.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22791217
CAS Number: 131030-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR