5-Bromo-2-[(1,1-dimethylethyl)thio]pyridine ≥97%

Supplier: AOBChem USA

Warning

167540-1G 167540-500MG 167540-5G 167540-25G
77465-658EA 236.9 USD
77465-658 77478-414 77488-602 77499-172
5-Bromo-2-[(1,1-dimethylethyl)thio]pyridine ≥97%
5-Bromo-2-[(1,1-dimethylethyl)thio]pyridine

Precaución: For research use only

Formula: C₉H₁₂BrNS
MW: 246.1672 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14652070
CAS Number: 123792-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR