Ethyl 4-chloro-2-fluoro-α-hydroxybenzeneacetate ≥95%

Supplier: AOBChem USA

Warning

167445-1G 167445-500MG 167445-5G
77465-654EA 546.93 USD
77465-654 77478-410 77488-598
Ethyl 4-chloro-2-fluoro-α-hydroxybenzeneacetate ≥95%
Ethyl 4-chloro-2-fluoro-α-hydroxybenzeneacetate

Precaución: For research use only

Formula: C₁₀H₁₀ClFO₃
MW: 232.64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23852821
CAS Number: 1526956-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR