3-Fluoro-2-[(phenylmethyl)oxy]benzoic acid ≥95%

Supplier: AOBChem USA

Warning

16737-1G 16737-500MG 16737-5G 16737-25G
77465-650EA 219.39 USD
77465-650 77478-406 77488-594 77499-170
3-Fluoro-2-[(phenylmethyl)oxy]benzoic acid ≥95%
3-Fluoro-2-[(phenylmethyl)oxy]benzoic acid

Precaución: For research use only

Formula: C₁₄H₁₁FO₃
MW: 246.234 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17372111
CAS Number: 938181-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR