Ethyl 3-bromo-2-fluoro-α-hydroxybenzeneacetate ≥95%

Supplier: AOBChem USA

Warning

167328-1G 167328-500MG 167328-5G 167328-25G
77465-646EA 412 USD
77465-646 77478-402 77488-590 77499-166
Ethyl 3-bromo-2-fluoro-α-hydroxybenzeneacetate ≥95%
Ethyl 3-bromo-2-fluoro-α-hydroxybenzeneacetate

Precaución: For research use only

Formula: C₁₀H₁₀BrFO₃
Storage Temperature: Ambient
MDL Number: MFCD34852185
CAS Number: 1501537-43-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR