Ethyl (4-bromophenyl)(hydroxy)acetate ≥96%

Supplier: AOBChem USA

Warning

167278-1G 167278-500MG 167278-5G 167278-25G
77465-642EA 246.17 USD
77465-642 77478-398 77488-586 77499-164
Ethyl (4-bromophenyl)(hydroxy)acetate ≥96%
Ethyl (4-bromophenyl)(hydroxy)acetate

Precaución: For research use only

Formula: C₁₀H₁₁BrO₃
Storage Temperature: Ambient
MDL Number: MFCD16693957
CAS Number: 30565-53-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR