1-chloro-2-fluoro-3-iodo-4-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

167086-1G 167086-500MG 167086-5G
77465-638EA 646.84 USD
77465-638 77478-394 77488-580
1-chloro-2-fluoro-3-iodo-4-methylbenzene ≥95%
1-chloro-2-fluoro-3-iodo-4-methylbenzene

Precaución: For research use only

Formula: C₇H₅ClFI
MW: 270.4704 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28751671
CAS Number: 1805523-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR