1-(3-acetylphenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

167076-1G 167076-500MG 167076-5G 167076-25G
77465-634EA 306.94 USD
77465-634 77478-390 77488-576 77499-156
1-(3-acetylphenyl)-2,2,2-trifluoroethanone ≥95%
1-(3-acetylphenyl)-2,2,2-trifluoroethanone

Precaución: For research use only

Formula: C₁₀H₇F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD13173346
CAS Number: 60283-15-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR