α-Ethyl-3-fluoro-4-methylbenzenemethanol ≥96%

Supplier: AOBChem USA

Warning

167067-1G 167067-500MG 167067-5G 167067-25G
77465-630EA 355.35 USD
77465-630 77478-386 77488-572 77499-152
α-Ethyl-3-fluoro-4-methylbenzenemethanol ≥96%
α-Ethyl-3-fluoro-4-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₀H₁₃FO
MW: 168.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11521206
CAS Number: 1183484-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR