α-Ethyl-4-fluoro-2-methylbenzenemethanol ≥96%

Supplier: AOBChem USA

Warning

167061-1G 167061-500MG 167061-5G
77465-626EA 471.74 USD
77465-626 77478-382 77488-568
α-Ethyl-4-fluoro-2-methylbenzenemethanol ≥96%
α-Ethyl-4-fluoro-2-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₀H₁₃FO
MW: 168.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11521260
CAS Number: 1270582-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR