2-Ethoxy-α-ethyl-5-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

167055-1G 167055-500MG 167055-5G
77465-622EA 546.93 USD
77465-622 77478-378 77488-564
2-Ethoxy-α-ethyl-5-methylbenzenemethanol ≥95%
2-Ethoxy-α-ethyl-5-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD16731926
CAS Number: 1285297-55-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR