α,3-Diethylbenzenemethanol ≥96%

Supplier: AOBChem USA

Warning

167053-1G 167053-500MG 167053-5G
77465-618EA 648.9 USD
77465-618 77478-374 77488-560
α,3-Diethylbenzenemethanol ≥96%
α,3-Diethylbenzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD11521244
CAS Number: 1314904-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR