3-(1,1-Dimethylethyl)-α-ethylbenzenemethanol ≥96%

Supplier: AOBChem USA

Warning

167050-1G 167050-500MG 167050-5G
77465-614EA 648.9 USD
77465-614 77478-370 77488-556
3-(1,1-Dimethylethyl)-α-ethylbenzenemethanol ≥96%
3-(1,1-Dimethylethyl)-α-ethylbenzenemethanol

Precaución: For research use only

Formula: C₁₃H₂₀O
Storage Temperature: Ambient
MDL Number: MFCD11521238
CAS Number: 1314961-90-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR