α-Ethyl-3-(methylthio)benzenemethanol ≥96%

Supplier: AOBChem USA

Warning

167041-1G 167041-500MG 167041-5G
77465-610EA 648.9 USD
77465-610 77478-366 77488-552
α-Ethyl-3-(methylthio)benzenemethanol ≥96%
α-Ethyl-3-(methylthio)benzenemethanol

Precaución: For research use only

Formula: C₁₀H₁₄OS
MW: 182.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11656010
CAS Number: 1378820-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR