α-Ethyl-2,6-difluoro-3-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

167027-1G 167027-500MG 167027-5G 167027-25G
77465-606EA 415.09 USD
77465-606 77478-362 77488-548 77499-146
α-Ethyl-2,6-difluoro-3-methylbenzenemethanol ≥95%
α-Ethyl-2,6-difluoro-3-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₀H₁₂F₂O
Storage Temperature: Ambient
MDL Number: MFCD24310363
CAS Number: 1545919-17-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR