3-Bromo-α-ethyl-2-fluoro-5-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

167004-1G 167004-500MG 167004-5G
77465-602EA 447.02 USD
77465-602 77478-358 77488-544
3-Bromo-α-ethyl-2-fluoro-5-methylbenzenemethanol ≥95%
3-Bromo-α-ethyl-2-fluoro-5-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₀H₁₂BrFO
MW: 247.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34852114
CAS Number: 2299718-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR