α-Ethyl-4-propylbenzenemethanol ≥96%

Supplier: AOBChem USA

Warning

166979-1G 166979-500MG 166979-5G
77465-598EA 648.9 USD
77465-598 77478-354 77488-540
α-Ethyl-4-propylbenzenemethanol ≥96%
α-Ethyl-4-propylbenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₈O
Storage Temperature: Ambient
MDL Number: MFCD11521232
CAS Number: 1215975-15-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR