α-Ethyl-3,5-dimethylbenzenemethanol ≥96%

Supplier: AOBChem USA

Warning

166963-1G 166963-500MG 166963-5G 166963-25G
77465-594EA 335.78 USD
77465-594 77478-350 77488-536 77499-140
α-Ethyl-3,5-dimethylbenzenemethanol ≥96%
α-Ethyl-3,5-dimethylbenzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD11521233
CAS Number: 1183794-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR