2-Chloro-α-ethyl-6-fluorobenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

166958-1G 166958-500MG 166958-5G
77465-590EA 493.37 USD
77465-590 77478-346 77488-532
2-Chloro-α-ethyl-6-fluorobenzenemethanol ≥95%
2-Chloro-α-ethyl-6-fluorobenzenemethanol

Precaución: For research use only

Formula: C₉H₁₀ClFO
MW: 188.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16302801
CAS Number: 1226405-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR