α-Ethyl-2,5-dimethylbenzenemethanol ≥96%

Supplier: AOBChem USA

Warning

166941-1G 166941-500MG 166941-5G 166941-25G
77465-586EA 382.13 USD
77465-586 77478-342 77488-528 77499-136
α-Ethyl-2,5-dimethylbenzenemethanol ≥96%
α-Ethyl-2,5-dimethylbenzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD11521255
CAS Number: 104174-12-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR