α-Ethyl-2,5-dimethoxybenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

166938-1G 166938-500MG 166938-5G 166938-25G
77465-582EA 341.96 USD
77465-582 77478-338 77488-524 77499-132
α-Ethyl-2,5-dimethoxybenzenemethanol ≥95%
α-Ethyl-2,5-dimethoxybenzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD11521223
CAS Number: 89106-42-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR