α-Ethyl[1,1'-biphenyl]-4-methanol ≥95%

Supplier: AOBChem USA

Warning

166927-1G 166927-500MG 166927-5G 166927-25G
77465-578EA 335.78 USD
77465-578 77478-334 77488-518 77499-126
α-Ethyl[1,1'-biphenyl]-4-methanol ≥95%
α-Ethyl[1,1'-biphenyl]-4-methanol

Precaución: For research use only

Formula: C₁₅H₁₆O
MW: 212.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11617678
CAS Number: 87292-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR