α-Ethyl-1-naphthalenemethanol ≥95%

Supplier: AOBChem USA

Warning

166917-1G 166917-500MG 166917-5G 166917-25G
77465-574EA 317.24 USD
77465-574 77478-330 77488-514 77499-122
α-Ethyl-1-naphthalenemethanol ≥95%
α-Ethyl-1-naphthalenemethanol

Precaución: For research use only

Formula: C₁₃H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD11521205
CAS Number: 56620-47-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR