α-Ethyl-4-(ethylthio)-α-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

166868-1G 166868-500MG 166868-5G 166868-25G
77465-570EA 341.96 USD
77465-570 77478-326 77488-510 77499-118
α-Ethyl-4-(ethylthio)-α-methylbenzenemethanol ≥95%
α-Ethyl-4-(ethylthio)-α-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₈OS
MW: 210.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20528397
CAS Number: 1378874-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR