3-Ethoxy-α-ethyl-α-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

166847-1G 166847-500MG 166847-5G
77465-566EA 648.9 USD
77465-566 77478-322 77488-506
3-Ethoxy-α-ethyl-α-methylbenzenemethanol ≥95%
3-Ethoxy-α-ethyl-α-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD20528422
CAS Number: 1379366-96-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR