4-Butyl-α-ethyl-α-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

166792-1G 166792-500MG 166792-5G
77465-562EA 648.9 USD
77465-562 77478-318 77488-502
4-Butyl-α-ethyl-α-methylbenzenemethanol ≥95%
4-Butyl-α-ethyl-α-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₄H₂₂O
Storage Temperature: Ambient
MDL Number: MFCD20526615
CAS Number: 152300-86-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR