α-Ethyl-α-methyl-1-naphthalenemethanol ≥95%

Supplier: AOBChem USA

Warning

166778-1G 166778-500MG 166778-5G
77465-558EA 648.9 USD
77465-558 77478-314 77488-498
α-Ethyl-α-methyl-1-naphthalenemethanol ≥95%
α-Ethyl-α-methyl-1-naphthalenemethanol

Precaución: For research use only

Formula: C₁₄H₁₆O
MW: 200.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00197600
CAS Number: 102238-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR