4-(Propylthio)-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

166722-1G 166722-500MG 166722-5G 166722-25G
77465-554EA 415.09 USD
77465-554 77478-310 77488-494 77499-112
4-(Propylthio)-1,1'-biphenyl ≥95%
4-(Propylthio)-1,1'-biphenyl

Precaución: For research use only

Formula: C₁₅H₁₆S
MW: 228.35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34852036
CAS Number: 1869045-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR