1-Methoxy-4-(propylthio)benzene ≥95%

Supplier: AOBChem USA

Warning

166716-1G 166716-500MG 166716-5G 166716-25G
77465-550EA 134.93 USD
77465-550 77478-306 77488-490 77499-108
1-Methoxy-4-(propylthio)benzene ≥95%
1-Methoxy-4-(propylthio)benzene

Precaución: For research use only

Formula: C₁₀H₁₄OS
MW: 182.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26958718
CAS Number: 62291-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR