2,2-Difluoro-4-iodo-1,3-benzodioxole ≥95%

Supplier: AOBChem USA

Warning

166520-1G 166520-5G 166520-25G
77465-546EA 60.77 USD
77465-546 77488-484 77499-102
2,2-Difluoro-4-iodo-1,3-benzodioxole ≥95%
2,2-Difluoro-4-iodo-1,3-benzodioxole

Precaución: For research use only

Formula: C₇H₃F₂IO₂
Storage Temperature: Ambient
MDL Number: MFCD17016040
CAS Number: 531508-54-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR